missing translation for 'onlineSavingsMsg'
Learn More

2'-Fucosyllactose (~90%), TRC

Product Code. 30133278
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133278 2 mg 2mg
30136368 1 mg 1mg
30006397 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30133278 Supplier Toronto Research Chemicals Supplier No. TRCF8361382MG
 Restricted Item
Estimated Shipment: 29-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41263-94-9
Molecular Formula C18H32O15
Molecular Weight (g/mol) 488.44
Synonym Lactose, 2'-O-fucosyl- (7CI), O-6-Deoxy-α-L-galactopyranosyl-(1→2)-O-β-D-galactopyranosyl-(1→4)-D-glucose, 2'-Fucosyllactose, 2'-O-Fucosyllactose
SMILES C[C@@H]1O[C@@H](O[C@@H]2[C@@H](O)[C@@H](O)[C@@H](CO)O[C@H]2O[C@H]([C@H](O)CO)[C@H](O)[C@@H](O)C=O)[C@@H](O)[C@H](O)[C@@H]1O

Specifications

Quantity 2 mg
InChI Formula InChI=1S/C18H32O15/c1-5-9(24)12(27)14(29)17(30-5)33-16-13(28)11(26)8(4-21)31-18(16)32-15(7(23)3-20)10(25)6(22)2-19/h2,5-18,20-29H,3-4H2,1H3/t5-,6-,7+,8+,9+,10+,11-,12+,13-,14-,15+,16+,17-,18-/m0/s1
Formula Weight 488.1741
Percent Purity >95%
Chemical Name or Material 2'-Fucosyllactose
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.