missing translation for 'onlineSavingsMsg'
Learn More

2-Formylthiophene-4-boronic acid, 98%, Thermo Scientific™

Product Code. 15480038
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15480038 1 g 1g
15490038 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15480038 Supplier Thermo Scientific Alfa Aesar Supplier No. H53298.03
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
2-Formylthiophene-4-boronic acid is used as intermediates.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, heat. Refrigerate.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175592-59-3
Molecular Formula C5H5BO3S
Molecular Weight (g/mol) 155.962
MDL Number MFCD03002365
InChI Key ZCUFJAORCNFWOF-UHFFFAOYSA-N
Synonym 2-formylthiophene-4-boronic acid, 5-formylthiophen-3-yl boronic acid, 5-formylthiophene-3-boronic acid, 5-formyl-3-thienyl boronic acid, 4-borono-2-formylthiophene, 5-formylthiophen-3-boronic acid, 5-formyl-3-thienylboronic acid, 5-formyl-3-thiopheneboronic acid, 4-boronothiophene-2-carboxaldehyde, 2-formyl-4-thiopheneboronic acid
PubChem CID 11084115
IUPAC Name (5-formylthiophen-3-yl)boronic acid
SMILES B(C1=CSC(=C1)C=O)(O)O

Specifications

Melting Point 113°C
Quantity 1 g
Solubility Information Slightly soluble in water.
Formula Weight 156.97
Percent Purity 98%
Chemical Name or Material 2-Formylthiophene-4-boronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.