missing translation for 'onlineSavingsMsg'
Learn More

2-Formylphenylboronic acid, 98%

Product Code. 10733783 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10733783 5 g Glass Bottle 5g
10272702 1 g Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10733783 Supplier Thermo Scientific Acros Supplier No. 359360050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40138-16-7 | C7H7BO3 | 149.94 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40138-16-7
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151822
InChI Key DGUWACLYDSWXRZ-UHFFFAOYSA-N
Synonym 2-formylphenyl boronic acid, 2-formylbenzeneboronic acid, benzaldehyde-2-boronic acid, o-formylphenylboronic acid, 2-boronobenzaldehyde, 2-dihydroxyboryl benzaldehyde, boronic acid, 2-formylphenyl, 2-benzaldehyde boronic acid, 2-formylphenyl-boronic acid
PubChem CID 292189
IUPAC Name (2-formylphenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1C=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 120.0°C to 122.0°C
Color Orange-Pink to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 149.94
Percent Purity 98%
Physical Form Crystalline Powder, Needles or Chunks
Chemical Name or Material 2-Formylphenylboronic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Formylphenylboronic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 2-Formylphenylboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.