missing translation for 'onlineSavingsMsg'
Learn More

2-Formylbenzeneboronic acid, 97%

Product Code. 11447116
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11447116 5 g 5g
11437116 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11447116 Supplier Thermo Scientific Alfa Aesar Supplier No. B25434.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 40138-16-7 | C7H7BO3 | 149.94 g/mol

2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.

Solubility
Soluble in methanol.

Notes
Air sensitive. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40138-16-7
Molecular Formula C7H7BO3
Molecular Weight (g/mol) 149.94
MDL Number MFCD00151822
InChI Key DGUWACLYDSWXRZ-UHFFFAOYSA-N
Synonym 2-formylphenyl boronic acid, 2-formylbenzeneboronic acid, benzaldehyde-2-boronic acid, o-formylphenylboronic acid, 2-boronobenzaldehyde, 2-dihydroxyboryl benzaldehyde, boronic acid, 2-formylphenyl, 2-benzaldehyde boronic acid, 2-formylphenyl-boronic acid
PubChem CID 292189
IUPAC Name (2-formylphenyl)boronic acid
SMILES OB(O)C1=CC=CC=C1C=O

Specifications

Melting Point ∼115°C to 120°C
Quantity 5 g
Beilstein 3030776
Sensitivity Air Sensitive
Solubility Information Soluble in methanol.
Formula Weight 149.94
Percent Purity 97%
Chemical Name or Material 2-Formylbenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.