Learn More
2-Formylbenzeneboronic acid, 97%
Description
2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Formylbenzeneboronic acid is used as an organic chemical synthesis intermediate.
Solubility
Soluble in methanol.
Notes
Air sensitive. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | ∼115°C to 120°C |
| Quantity | 1 g |
| Beilstein | 3030776 |
| Sensitivity | Air Sensitive |
| Solubility Information | Soluble in methanol. |
| Formula Weight | 149.94 |
| Percent Purity | 97% |
| Chemical Name or Material | 2-Formylbenzeneboronic acid |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Politique de confidentialité.