missing translation for 'onlineSavingsMsg'
Learn More

2-Formyl-3-thiopheneboronic acid, 97%

Product Code. 10461420 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10461420 5 g Glass bottle 5g
10765252 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10461420 Supplier Thermo Scientific Acros Supplier No. 427690050
 Restricted Item
Estimated Shipment: 02-02-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4347-31-3 | C5H5BO3S | 155.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4347-31-3
Molecular Formula C5H5BO3S
Molecular Weight (g/mol) 155.96
MDL Number MFCD01075678
InChI Key BBENFHSYKBYWJX-UHFFFAOYSA-N
Synonym 2-formylthiophene-3-boronic acid, 2-formylthiophen-3-yl boronic acid, 2-formyl-3-thiopheneboronic acid, 3-borono-2-formylthiophene, 2-formyl-3-thienyl boronic acid, 2-formyl-3-thienylboronic acid, 3-boronothiophene-2-carboxaldehyde, 2-formylthien-3-ylboronic acid, boronic acid, 2-formyl-3-thienyl, 2-formylthiophene-3-boronicacid
PubChem CID 2773426
IUPAC Name (2-formylthiophen-3-yl)boronic acid
SMILES OB(O)C1=C(SC=C1)C=O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 137.0°C to 193.0°C
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 155.99
Percent Purity 97%
Chemical Name or Material 2-Formyl-3-thiopheneboronic acid
compliance-icons
Product Identifier
  • 2-Formyl-3-thiopheneboronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.