missing translation for 'onlineSavingsMsg'
Learn More

2-(Fmoc-amino)benzoic acid, 98%, Thermo Scientific Chemicals

Product Code. 15440169
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field

Solubility
Soluble in water or 1% acetic acid.

Notes
Refrigerate. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150256-42-1
Molecular Formula C22H17NO4
Molecular Weight (g/mol) 359.381
MDL Number MFCD00235883
InChI Key CNAVPEPPAVHHKN-UHFFFAOYSA-N
Synonym fmoc-2-abz-oh, fmoc-2-aminobenzoic acid, 2-fmoc-amino benzoic acid, fmoc-abz-oh, n-fmoc-anthranilic acid, 2-9h-fluoren-9-yl methoxy carbonyl amino benzoic acid, fmoc-anthranilic acid, benzoic acid, 2-9h-fluoren-9-ylmethoxy carbonyl amino, 2-9h-fluoren-9-ylmethoxy carbonyl amino benzoic acid, n-2-9-fluorenylmethyloxycarbonyl amino-benzoic acid
PubChem CID 978386
IUPAC Name 2-(9H-fluoren-9-ylmethoxycarbonylamino)benzoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC4=CC=CC=C4C(=O)O

Specifications

Melting Point 216°C to 218°C (decomposition)
Color White
Quantity 25 g
Beilstein 7659726
Solubility Information Soluble in water or 1% acetic acid.
Formula Weight 359.37
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-(Fmoc-amino)benzoic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'