missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine, 99%

Product Code. 11454723
Change view
Click to view available options
Quantity:
10 g
25 g
100 g
250 g
Unit Size:
100g
10g
250g
25g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11454723 250 g 250g
11444723 10 g 10g
11389609 25 g 25g
11399609 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 11454723 Supplier Thermo Scientific Alfa Aesar Supplier No. A11627.30
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 372-48-5 | C5H4FN | 97.092 g/mol

2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoropyridine is a useful precursor of 2,3-disubstituted pyridines which is made by the possibility of metallation at C-3, together with the ready nucleophilic displacement of the activated fluorine. It can also react with thiophene to get 2-thiophen-2-yl-pyridine. It is used in the synthesis of aminopyridines.

Solubility
Soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 372-48-5
Molecular Formula C5H4FN
Molecular Weight (g/mol) 97.092
MDL Number MFCD00006224
InChI Key MTAODLNXWYIKSO-UHFFFAOYSA-N
Synonym pyridine, 2-fluoro, o-fluoropyridine, 2-fluoro-pyridine, pyridine, fluoro, ccris 1725, 2-fluoranylpyridine, fluoropyridine, 2fluoropyridine, 2-fluorpyridin, 2-fluoro-pyridinium
PubChem CID 9746
IUPAC Name 2-fluoropyridine
SMILES C1=CC=NC(=C1)F

Specifications

Density 1.129
Boiling Point 124°C to 126°C
Flash Point 24°C (75°F)
Refractive Index 1.467
Quantity 250 g
UN Number UN1993
Beilstein 1515
Solubility Information Soluble in water.
Formula Weight 97.09
Percent Purity 99%
Chemical Name or Material 2-Fluoropyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.