missing translation for 'onlineSavingsMsg'
Learn More

6-Fluoro-3-pyridinylboronic acid, 97%, Thermo Scientific™

Catalog Number 10031304
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
351019-18-6
C5H5BFNO2
140.91
MFCD03411559
OJBYZWHAPXIJID-UHFFFAOYSA-N
2-fluoropyridine-5-boronic acid, 2-fluoro-5-pyridylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridine-3-boronic acid, 2-fluoro-5-pyridineboronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridin-5-boronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoro-3-pyridineboronic acid
2783397
(6-fluoropyridin-3-yl)boronic acid
OB(O)C1=CC=C(F)N=C1
This item is not returnable. View return policy
351019-18-6
C5H5BFNO2
140.91
MFCD03411559
OJBYZWHAPXIJID-UHFFFAOYSA-N
2-fluoropyridine-5-boronic acid, 2-fluoro-5-pyridylboronic acid, 6-fluoropyridin-3-yl boronic acid, 6-fluoropyridine-3-boronic acid, 2-fluoro-5-pyridineboronic acid, 2-fluoropyridin-5-ylboronic acid, 2-fluoropyridin-5-boronic acid, 6-fluoro-3-pyridinylboronic acid, 6-fluoropyridin-3-yl-3-boronic acid, 6-fluoro-3-pyridineboronic acid
2783397
(6-fluoropyridin-3-yl)boronic acid
OB(O)C1=CC=C(F)N=C1
CAS Min % 96.0
CAS Max % 100.0
Melting Point 172.0°C to 176.0°C
Assay Percent Range 97%
Packaging Glass bottle
Quantity 1 g
Formula Weight 140.91
Percent Purity 97%
Chemical Name or Material 6-Fluoro-3-pyridinylboronic acid
compliance-icons
Product Identifier
  • 2-Fluoropyridine-5-boronic acid
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'