missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine-4-methanol, 97%, Thermo Scientific Chemicals

Product Code. 15279766
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131747-60-9
Molecular Formula C6H6FNO
Molecular Weight (g/mol) 127.12
InChI Key IAFXOPAZVVMMLE-UHFFFAOYSA-N
Synonym 2-fluoropyridin-4-yl methanol, 2-fluoro-4-pyridinemethanol, 2-fluoro-pyridin-4-yl-methanol, 4-pyridinemethanol, 2-fluoro, acmc-1c3lv, 2-fluoropyridine-4-methanol, 2-fluoro-4-hydroxymethylpyridine, 4-pyridinemethanol, 2-fluoro-9ci
PubChem CID 11332468
IUPAC Name (2-fluoropyridin-4-yl)methanol
SMILES C1=CN=C(C=C1CO)F

Specifications

Melting Point 61°C to 63°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 127.12
Percent Purity 97%
Chemical Name or Material 2-Fluoropyridine-4-methanol
compliance-icons
Product Identifier
  • 2-Fluoropyridine-4-methanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'