missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyridine-4-boronic acid pinacol ester, 95%

Product Code. 11476330
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11476330 5 g 5g
11466330 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476330 Supplier Thermo Scientific Alfa Aesar Supplier No. H27179.06
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 458532-86-0 | C11H15BFNO2 | 223.05 g/mol

Used in pharmaceuticals, agrochemicals, and high-tech materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in pharmaceuticals, agrochemicals, and high-tech materials.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 458532-86-0
Molecular Formula C11H15BFNO2
Molecular Weight (g/mol) 223.05
MDL Number MFCD06798253
InChI Key PCLMNCBIXQQRMB-UHFFFAOYSA-N
Synonym 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridine-4-boronic acid pinacol ester, 2-fluoro-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-fluoropyridin-4-boronic acid pinacol ester, 2-fluoropyridin-4-ylboronic acid pinacol ester, pyridine, 2-fluoro-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, abbypharma ap-11-5396, 2-fluoro-4-pyridineboronic acid pinacol ester, 2-fluoropyridin-4-yl boronic acid pinacol ester, 2-2-fluoro 4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
PubChem CID 12060164
IUPAC Name 2-fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CC(F)=NC=C1

Specifications

Melting Point 48°C to 52°C
Quantity 5 g
Formula Weight 223.06
Percent Purity 95%
Chemical Name or Material 2-Fluoropyridine-4-boronic acid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.