missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoropyrazine, 98%, Thermo Scientific™

Product Code. 15515946 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15515946 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15515946 Supplier Thermo Scientific Acros Supplier No. 456960010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 4949-13-7
Density 1.2130g/mL
Boiling Point 110.0°C
Flash Point 21°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Molecular Formula C4H3FN2
MDL Number MFCD03094084
Quantity 1 g
Specific Gravity 1.213
Synonym 2-fluoro-pyrazine, fluoropyrazine, pyrazine, fluoro, pyrazine, 2-fluoro, 2-fluoranylpyrazine, pubchem13961, 2-fluoro-1,4-diazine
InChI Key IJUHIGIZXFLJOY-UHFFFAOYSA-N
SMILES FC1=NC=CN=C1
IUPAC Name 2-fluoropyrazine
Molecular Weight (g/mol) 98.08
PubChem CID 642835
Formula Weight 98.08
Percent Purity 98%
Chemical Name or Material 2-Fluoropyrazine
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Fluoropyrazine, 98%
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H318-Causes serious eye damage.
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.