missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylacetonitrile, 97%

Product Code. 11434703
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11434703 5 g 5g
11444703 25 g 25g
11454703 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11434703 Supplier Thermo Scientific Alfa Aesar Supplier No. A11612.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 326-62-5 | C8H6FN | 135.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 326-62-5
Molecular Formula C8H6FN
Molecular Weight (g/mol) 135.14
MDL Number MFCD00001897
InChI Key DAVJMKMVLKOQQC-UHFFFAOYSA-N
Synonym 2-fluorophenylacetonitrile, 2-fluorobenzyl cyanide, 2-2-fluorophenyl acetonitrile, benzeneacetonitrile, 2-fluoro, o-fluorophenylacetonitrile, o-fluorobenzyl cyanide, 2-fluorophenyl acetonitrile, acetonitrile, o-fluorophenyl, 2-2-fluorophenyl ethanenitrile, 2-fluorobenzeneacetonitrile
PubChem CID 67592
IUPAC Name 2-(2-fluorophenyl)acetonitrile
SMILES FC1=CC=CC=C1CC#N

Specifications

Density 1.061
Boiling Point 235°C
Flash Point >100°C (212°F)
Refractive Index 1.501
Quantity 5 g
UN Number UN3276
Beilstein 1862361
Formula Weight 135.14
Percent Purity 97%
Chemical Name or Material 2-Fluorophenylacetonitrile

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.