missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenylacetic acid, 98+%

Product Code. 11403274
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11403274 10 g 10g
11413274 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11403274 Supplier Thermo Scientific Alfa Aesar Supplier No. A10596.09
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 451-82-1 | C8H7FO2 | 154.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 451-82-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD00004315
InChI Key RPTRFSADOICSSK-UHFFFAOYSA-N
Synonym 2-fluorophenylacetic acid, 2-2-fluorophenyl acetic acid, o-fluorophenylacetic acid, benzeneacetic acid, 2-fluoro, 2-fluorophenyl acetic acid, 2-fluorophenylaceticacid, acetic acid, o-fluorophenyl, pubchem4164, 2fluorophenylacetic acid
PubChem CID 67979
IUPAC Name 2-(2-fluorophenyl)acetic acid
SMILES C1=CC=C(C(=C1)CC(=O)O)F

Specifications

Melting Point 61°C to 65°C
Odor Odorless
Quantity 10 g
Beilstein 775955
Formula Weight 154.14
Percent Purity ≥98%
Chemical Name or Material 2-Fluorophenylacetic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.