missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenyl isocyanate, 98%

Product Code. 11428067
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11428067

Brand: Thermo Scientific Alfa Aesar A17466.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 16744-98-2 | C7H4FNO | 137.113 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 16744-98-2
Molecular Formula C7H4FNO
Molecular Weight (g/mol) 137.113
MDL Number MFCD00001996
InChI Key VZNCSZQPNIEEMN-UHFFFAOYSA-N
Synonym 2-fluorophenyl isocyanate, o-fluorophenyl isocyanate, 2-fluorophenylisocyanate, benzene, 1-fluoro-2-isocyanato, isocyanic acid 2-fluorophenyl ester, 2-fluorobenzenisocyanate, paragos 331045, labotest-bb ltbb005297, pubchem4455, 2-flurophenylisocyanate
PubChem CID 85588
IUPAC Name 1-fluoro-2-isocyanatobenzene
SMILES C1=CC=C(C(=C1)N=C=O)F

Specifications

Density 1.221
Boiling Point 56°C to 58°C (10 mmHg)
Flash Point 50°C (122°F)
Odor Pungent, Makes Eyes Water
Refractive Index 1.5125
Quantity 25 g
UN Number UN3080
Beilstein 1210127
Sensitivity Moisture sensitive
Formula Weight 137.11
Percent Purity 98%
Chemical Name or Material 2-Fluorophenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.