missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenyl isocyanate, ≥95%, Maybridge

Product Code. 10667113
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10667113 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10667113 Supplier Thermo Scientific Maybridge Supplier No. SB01843DA

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 16744-98-2
CAS Min % 95.0
Density 1.222g/mL
Assay Percent Range 95%+
Molecular Formula C7H4FNO
Linear Formula FC6H4NCO
MDL Number MFCD00001996
Quantity 1 g
Synonym 2-fluorophenyl isocyanate, o-fluorophenyl isocyanate, 2-fluorophenylisocyanate, benzene, 1-fluoro-2-isocyanato, isocyanic acid 2-fluorophenyl ester, 2-fluorobenzenisocyanate, paragos 331045, labotest-bb ltbb005297, pubchem4455, 2-flurophenylisocyanate
InChI Key VZNCSZQPNIEEMN-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)N=C=O)F
IUPAC Name 1-fluoro-2-isocyanatobenzene
Molecular Weight (g/mol) 137.113
PubChem CID 85588
Formula Weight 137.11
Percent Purity 95%
Chemical Name or Material 2-Fluorophenyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.