missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenol, 98%

Product Code. 11487187
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11487187 100 g 100g
11477187 25 g 25g
11497187 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11487187 Supplier Thermo Scientific Alfa Aesar Supplier No. A14979.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 367-12-4 | C6H5FO | 112.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-12-4
Molecular Formula C6H5FO
Molecular Weight (g/mol) 112.10
MDL Number MFCD00002155
InChI Key HFHFGHLXUCOHLN-UHFFFAOYSA-N
Synonym o-fluorophenol, phenol, 2-fluoro, phenol, o-fluoro, 1-fluoro-2-hydroxybenzene, 2-fluorohydroxybenzene, fluorophenol, unii-i9ow1nly9r, 2-fluoro-phenol, phenol, fluoro, ccris 1226
PubChem CID 9707
IUPAC Name 2-fluorophenol
SMILES OC1=CC=CC=C1F

Specifications

Melting Point 14°C to 16°C
Density 1.215
Boiling Point 150°C to 152°C
Flash Point 46°C (114°F)
Odor Phenol-like
Refractive Index 1.514
Quantity 100 g
UN Number UN2924
Beilstein 1905112
Formula Weight 112.1
Percent Purity 98%
Chemical Name or Material 2-Fluorophenol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.