missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorophenacyl bromide, 98%

Product Code. 12075400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
12075400 25 g 25g
12065400 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 12075400 Supplier Thermo Scientific Acros Supplier No. 446670250
 Restricted Item
Estimated Shipment: 16-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 655-15-2 | C8H6BrFO | 217.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 655-15-2
Molecular Formula C8H6BrFO
Molecular Weight (g/mol) 217.04
InChI Key QDNWNJSLWKHNTM-UHFFFAOYSA-N
Synonym 2-bromo-2'-fluoroacetophenone, 2-bromo-1-2-fluorophenyl ethanone, 2-fluorophenacyl bromide, 2-bromo-1-2-fluorophenyl ethan-1-one, alpha-bromo-2-fluoroacetophenone, 2-fluorophenacylbromide, ethanone, 2-bromo-1-2-fluorophenyl, o-fluorophenacyl bromide, omega-bromo-2-fluoroacetophenone, 2-bromo-1-2'-fluorophenyl ethanone
PubChem CID 2737449
IUPAC Name 2-bromo-1-(2-fluorophenyl)ethanone
SMILES C1=CC=C(C(=C1)C(=O)CBr)F

Specifications

Boiling Point 61.0°C to 63.0°C (0.1 Torr)
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 217.04
Percent Purity 98%
Chemical Name or Material 2-Fluorophenacyl bromide
compliance-icons compliance-icons
Product Identifier
  • 2-Fluorophenacyl bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.