missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoromesitylene, 98%

Product Code. 11425123
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11425123

Brand: Thermo Scientific Alfa Aesar A12779.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 392-69-8 | C9H11F | 138.19 g/mol

2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-69-8
Molecular Formula C9H11F
Molecular Weight (g/mol) 138.19
MDL Number MFCD00052365
InChI Key ZLGPNBBJPOBSLY-UHFFFAOYSA-N
Synonym fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939
PubChem CID 136234
IUPAC Name 2-fluoro-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(F)C(C)=C1

Specifications

Density 0.975
Boiling Point 163°C to 165°C
Refractive Index 1.483
Quantity 5 g
Solubility Information Soluble in water.
Formula Weight 138.19
Percent Purity 98%
Chemical Name or Material 2-Fluoromesitylene
compliance-icons
Product Identifier
  • 2,4,6-Trimethylfluorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Mesityl fluoride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.