missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoromesitylene, 98%

Product Code. 11415123
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11415123 1 g 1g
11425123 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11415123 Supplier Thermo Scientific Alfa Aesar Supplier No. A12779.03
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 392-69-8 | C9H11F | 138.19 g/mol

2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Fluoromesitylene, is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals and Dyestuff.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place.Stable under recommended storage conditions. Incompatible to strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 392-69-8
Molecular Formula C9H11F
Molecular Weight (g/mol) 138.19
MDL Number MFCD00052365
InChI Key ZLGPNBBJPOBSLY-UHFFFAOYSA-N
Synonym fluoromesitylene, mesityl fluoride, 2-fluoromesitylene, 2,4,6-trimethylfluorobenzene, benzene, 2-fluoro-1,3,5-trimethyl, 1-fluoro-2,4,6-trimethylbenzene, 2-fluoro-1,3,5-trimethyl-benzene, fluormesitylen, mesitylfluoride, pubchem16939
PubChem CID 136234
IUPAC Name 2-fluoro-1,3,5-trimethylbenzene
SMILES CC1=CC(C)=C(F)C(C)=C1

Specifications

Density 0.975
Boiling Point 163°C to 165°C
Refractive Index 1.483
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 138.19
Percent Purity 98%
Chemical Name or Material 2-Fluoromesitylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.