missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroethyl Chloroformate, TRC

Product Code. 30393605
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30393605 5 g
30406970 10 g
30403967 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393605 Supplier Toronto Research Chemicals Supplier No. TRCF5915305G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 462-27-1
Molecular Formula C3H4ClFO2
Molecular Weight (g/mol) 126.51
Synonym Carbonochloridic Acid 2-Fluoroethyl Ester, 2-Fluoroethyl Carbonochloridate
SMILES FCCOC(=O)Cl

Specifications

Quantity 5 g
InChI Formula InChI=1S/C3H4ClFO2/c4-3(6)7-2-1-5/h1-2H2
Chemical Name or Material 2-Fluoroethyl Chloroformate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.