missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroethanol, 95%

Product Code. 12674937 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12674937 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12674937 Supplier Thermo Scientific Acros Supplier No. 170830050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 371-62-0 | C2H5FO | 64.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 371-62-0
Molecular Formula C2H5FO
Molecular Weight (g/mol) 64.06
InChI Key GGDYAKVUZMZKRV-UHFFFAOYSA-N
Synonym ethanol, 2-fluoro, fluoroethanol, 2-fluoro-1-ethanol, ethylene fluorohydrin, beta-fluoroethanol, 2-fluoroethan-1-ol, monofluorethanol, monofluoroethanol, 2-fluoro-ethanol, unii-4m427a5hml
PubChem CID 9737
IUPAC Name 2-fluoroethanol
SMILES C(CF)O

Specifications

Melting Point -26.5°C
Density 1.1080g/mL
Boiling Point 100.0°C to 104.0°C
Flash Point 34°C
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula FCH2CH2OH
Quantity 5 g
Beilstein 01,I,17
Specific Gravity 1.108
Solubility Information Solubility in water: miscible
Formula Weight 64.06
Percent Purity 95%
Chemical Name or Material 2-Fluoroethanol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluoroethanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H330-Fatal if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.