missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzylamine, 96%

Product Code. 10751831 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10751831 10 g Glass bottle 10g
10522201 1 g Glass bottle 1g
10239633 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10751831 Supplier Thermo Scientific Acros Supplier No. 171940100
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 89-99-6 | C7H8FN | 125.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-99-6
Molecular Formula C7H8FN
Molecular Weight (g/mol) 125.15
MDL Number MFCD00008107
InChI Key LRFWYBZWRQWZIM-UHFFFAOYSA-N
Synonym 2-fluorobenzylamine, 2-fluorophenyl methanamine, benzenemethanamine, 2-fluoro, o-fluorobenzylamine, benzylamine, o-fluoro, 1-2-fluorophenyl methanamine, 2-fluoro-benzylamine, 2-fluorobenzyl amine, 2-fluorophenyl methylamine, o-fluorobenzyl amine
PubChem CID 66649
IUPAC Name (2-fluorophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)F

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.0900g/mL
Boiling Point 73.0°C to 75.0°C (13.0 mmHg)
Flash Point 67°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4CH2NH2
Refractive Index 1.5160 to 1.5190
Quantity 10 g
Specific Gravity 1.09
Formula Weight 125.15
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Fluorobenzylamine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.