missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzyl bromide, 98%

Product Code. 11490593
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11490593 10 g 10g
11400603 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11490593 Supplier Thermo Scientific Alfa Aesar Supplier No. A12692.09
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 446-48-0 | C7H6BrF | 189.027 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 446-48-0
Molecular Formula C7H6BrF
Molecular Weight (g/mol) 189.027
MDL Number MFCD00000324
InChI Key FFWQLZFIMNTUCZ-UHFFFAOYSA-N
Synonym 2-fluorobenzyl bromide, 1-bromomethyl-2-fluorobenzene, 2-fluorobenzylbromide, o-fluorobenzyl bromide, alpha-bromo-o-fluorotoluene, alpha-bromo-2-fluorotoluene, 2-fluoro-benzyl bromide, 1-bromomethyl-2-fluoro-benzene, benzene, 1-bromomethyl-2-fluoro, 2-fluoro-benzylbromide
PubChem CID 67968
IUPAC Name 1-(bromomethyl)-2-fluorobenzene
SMILES C1=CC=C(C(=C1)CBr)F

Specifications

Boiling Point 84°C to 85°C (15 mmHg)
Flash Point 82°C (179°F)
Odor Pungent
Quantity 10 g
UN Number UN3265
Beilstein 1099894
Formula Weight 189.03
Percent Purity 98%
Chemical Name or Material 2-Fluorobenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.