missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzotrifluoride, 99%, ACROS Organics™

Product Code. 10282552 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10282552 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10282552 Supplier Thermo Scientific Acros Supplier No. 204460050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 392-85-8
CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 1.2900g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Molecular Formula C7H4F4
Linear Formula FC6H4CF3
Refractive Index 1.4045 to 1.4065 (20°C, 589nm)
MDL Number MFCD00000374
Quantity 5 g
Synonym 2-fluorobenzotrifluoride, 1-fluoro-2-trifluoromethyl benzene, o-fluorobenzotrifluoride, benzene, 1-fluoro-2-trifluoromethyl, alpha,alpha,alpha,2-tetrafluorotoluene, benzene, fluoro trifluoromethyl, o,alpha,alpha,alpha-tetrafluorotoluene, a,a,a,2-tetrafluorotoluene, alpha,alpha,alpha,ar-tetrafluorotoluene, 2-trifluoromethyl fluorobenzene
InChI Key BGVGHYOIWIALFF-UHFFFAOYSA-N
SMILES C1=CC=C(C(=C1)C(F)(F)F)F
IUPAC Name 1-fluoro-2-(trifluoromethyl)benzene
Molecular Weight (g/mol) 164.103
PubChem CID 9807
Formula Weight 164.1
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material 2-Fluorobenzotrifluoride, 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.