missing translation for 'onlineSavingsMsg'
Learn More

2-Fluorobenzene-1-sulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10262832
Change view
Click to view available options
Quantity:
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10262832 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10262832 Supplier Thermo Scientific Maybridge Supplier No. KM01957DA
 Restricted Item
Estimated Shipment: 12-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 2905-21-7
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C6H4ClFO2S
Linear Formula FC6H4SO2Cl
MDL Number MFCD00042285
Quantity 1 g
Synonym 2-fluorobenzene-1-sulfonyl chloride, 2-fluorobenzenesulphonyl chloride, 2-fluorobenzenesulfonylchloride, 2-fluoro-benzenesulfonyl chloride, 2-fluorophenylsulfonyl chloride, benzenesulfonyl chloride, 2-fluoro, chloro 2-fluorophenyl sulfone, pubchem5138, ksc205g6f
InChI Key ZSZKAQCISWFDCQ-UHFFFAOYSA-N
SMILES FC1=CC=CC=C1S(Cl)(=O)=O
IUPAC Name 2-fluorobenzenesulfonyl chloride
Molecular Weight (g/mol) 194.60
PubChem CID 137761
Formula Weight 194.61
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzene-1-sulfonyl chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.