missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Fluorobenzal chloride, 97%

Product Code. 11376736
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11376736 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11376736 Supplier Alfa Aesar™ Supplier No. L03984.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 320-65-0
Molecular Formula C7H5Cl2F
Molecular Weight (g/mol) 179.02
MDL Number MFCD00043797
InChI Key XFAFBKQDEPXWCS-UHFFFAOYSA-N
Synonym 2-fluorobenzal chloride, 1-dichloromethyl-2-fluorobenzene, 2-fluorobenzalchloride, benzene, 1-dichloromethyl-2-fluoro, acmc-20anuj, 2-fluorobenzal chlorid, 2-fluoro-1-dichloromethylbenzene, toluene,.alpha.-dichloro-o-fluoro, alpha,alpha-dichloro-2-fluorotoluene, 1-dichloromethyl-2-fluoro-benzene
PubChem CID 67575
IUPAC Name 1-(dichloromethyl)-2-fluorobenzene
SMILES FC1=CC=CC=C1C(Cl)Cl

Specifications

Density 1.309
Boiling Point 224°C to 232°C
Refractive Index 1.527
Quantity 100 g
UN Number UN3265
Beilstein 2042648
Formula Weight 179.02
Percent Purity 97%
Chemical Name or Material 2-Fluorobenzal chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.