missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroanisole, 98%, Thermo Scientific™

Product Code. 11419533
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419533 100 g 100g
11409533 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11419533 Supplier Thermo Scientific Alfa Aesar Supplier No. A12380.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 321-28-8
Molecular Formula C7H7FO
Molecular Weight (g/mol) 126.13
MDL Number MFCD00000316
InChI Key JIXDOBAQOWOUPA-UHFFFAOYSA-N
Synonym 2-fluoroanisole, o-fluoroanisole, benzene, 1-fluoro-2-methoxy, ortho-fluoroanisole, anisole, o-fluoro, 1-fluoro-2-methoxy-benzene, paragos 390201, timtec-bb sbb040818, fr bo1 wln, fluoroanisole
PubChem CID 67576
IUPAC Name 1-fluoro-2-methoxybenzene
SMILES COC1=CC=CC=C1F

Specifications

Melting Point -39°C
Density 1.124
Boiling Point 154°C to 155°C
Flash Point 56°C (133°F)
Refractive Index 1.494
Quantity 100 g
UN Number UN1993
Beilstein 1859755
Formula Weight 126.13
Percent Purity 98%
Chemical Name or Material 2-Fluoroanisole
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.