missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroaniline, TRC

Product Code. 30390579
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30390579 50 g
30395263 10 g
30391983 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390579 Supplier Toronto Research Chemicals Supplier No. TRCF58828350G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-54-9
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
Synonym Benzenamine, 2-fluoro- (9CI, ACI), 2-Fluorobenzenamine (ACI), Aniline, o-fluoro- (8CI), 1-Amino-2-fluorobenzene, 2-Fluoroaniline, 2-Fluorophenylamine, NSC 10299, o-Fluoroaniline
IUPAC Name 2-fluoroaniline
SMILES Nc1ccccc1F

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H6FN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2
Formula Weight 111.0484
Chemical Name or Material 2-Fluoroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.