missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroaniline, 99+%

Product Code. 10437880 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10437880 100 mL Glass bottle 100mL
10714611 25 mL Glass bottle 25mL
10530584 500 mL Glass bottle 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10437880 Supplier Thermo Scientific Acros Supplier No. 119271000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 348-54-9 | C6H6FN | 111.12 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-54-9
Molecular Formula C6H6FN
Molecular Weight (g/mol) 111.12
MDL Number MFCD00007642
InChI Key FTZQXOJYPFINKJ-UHFFFAOYSA-N
Synonym o-fluoroaniline, benzenamine, 2-fluoro, 1-amino-2-fluorobenzene, 2-fluorobenzenamine, 2-fluoro-phenylamine, 2-fluoro-benzenamine, fluoroaniline-2, o-aminofluorobenzene, aniline, o-fluoro, 2-fluorophenylamine
PubChem CID 9584
ChEBI CHEBI:27526
IUPAC Name 2-fluoroaniline
SMILES NC1=CC=CC=C1F

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point -29°C
Color Colorless
Density 1.1510g/mL
Boiling Point 182°C to 183°C
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Linear Formula FC6H4NH2
Refractive Index 1.5400 to 1.542
Quantity 100 mL
Beilstein 12, I, 296
Specific Gravity 1.151
Solubility Information Solubility in water: 17g/L in water (20°C). Other solubilities: soluble in carbon tetrachloride,diethyl ether and,ethanol
Formula Weight 111.12
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Fluoroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.