missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoroadenosine, TRC

Product Code. 30401425
Change view
Click to view available options
Quantity:
100 mg
250 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30401425 500 mg
30396014 100 mg
30407221 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30401425 Supplier Toronto Research Chemicals Supplier No. TRCF587218500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 146-78-1
Molecular Formula C10H12FN5O4
Molecular Weight (g/mol) 285.23
Synonym 2-Fluoro-9-beta-D-ribofuranosyladenine, NSC 30605
SMILES Nc1nc(F)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C10H12FN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1
Formula Weight 285.0873
Chemical Name or Material 2-Fluoroadenosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.