missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals 2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine, 98%
Click to view available options
Quantity:
1 g
Unit Size:
1g
Description
2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine is used in drugs, cosmetics or foodstuffs.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2′-Fluoro-N2-isobutyryl-2′-deoxyguanosine is used in drugs, cosmetics or foodstuffs.
Solubility
Soluble in methanol
Notes
Protect from heat. Store away from oxidizing agents.
2′-Fluoro-N2-isobutyryl-2′-deoxyguanosine is used in drugs, cosmetics or foodstuffs.
Solubility
Soluble in methanol
Notes
Protect from heat. Store away from oxidizing agents.
Specifications
Specifications
| Chemical Name or Material | 2'-Fluoro-N2-isobutyryl-2'-deoxyguanosine |
| Color | White to Yellow |
| Molecular Formula | C14H18FN5O5 |
| MDL Number | MFCD15145246 |
| Quantity | 1 g |
| Solubility Information | Soluble in methanol |
| Formula Weight | 355.32 |
| Percent Purity | 98% |
| Physical Form | Powder |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction