missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15408736
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 313545-34-5
Molecular Formula C7H5BF4O2
Molecular Weight (g/mol) 207.919
MDL Number MFCD07781239
InChI Key TXBRJTKFAYXKMV-UHFFFAOYSA-N
Synonym 2-fluoro-6-trifluoromethyl phenylboronic acid, 2-fluoro-6-trifluoromethyl phenylboronicacid, 2-fluoro-6-trifluoromethyl benzeneboronic acid, 2-borono-3-fluorobenzotrifluoride, 2-fluoro-6-trifluoromethylphenylboronic acid, 2-fluoro-6-trifluoromethyl phenyl boronic acid, boronic acid, b-2-fluoro-6-trifluoromethyl phenyl, 2-fluoro-6-trifluoromethyl phenylboron, 2-fluoro-6-trifluoromethylphenyl boronic acid
PubChem CID 16217168
IUPAC Name [2-fluoro-6-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC=C1F)C(F)(F)F)(O)O

Specifications

Melting Point 115°C to 119°C
Quantity 5 g
Formula Weight 207.92
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 2-Fluoro-6-(trifluoromethyl)benzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'