missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-nitrobenzyl bromide, 98+%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Nitobenzyl ester based nonionic PAGs (NIPAG 2) NIPAG 2 was prepared from 2-fluoro-6-nitrobenzyl bromide with silver sulfonate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Nitobenzyl ester based nonionic PAGs (NIPAG 2) NIPAG 2 was prepared from 2-fluoro-6-nitrobenzyl bromide with silver sulfonate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Nitobenzyl ester based nonionic PAGs (NIPAG 2) NIPAG 2 was prepared from 2-fluoro-6-nitrobenzyl bromide with silver sulfonate.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, bases.
Specifications
Specifications
| Melting Point | 50°C to 54°C |
| Odor | Pungent |
| Quantity | 5 g |
| UN Number | UN3261 |
| Beilstein | 2503670 |
| Synonym | 2-fluoro-6-nitrobenzyl bromide, 2-bromomethyl-1-fluoro-3-nitrobenzene, benzene, 2-bromomethyl-1-fluoro-3-nitro, alpha-bromo-2-fluoro-6-nitrotoluene, acmc-20anuk, 2-fluoro-6-nitrobenzylbromide, 2-bromomethyl-3-fluoro-1-nitrobenzene, benzene,2-bromomethyl-1-fluoro-3-nitro |
| Solubility Information | Insoluble in water. |
| Formula Weight | 234.02 |
| Percent Purity | ≥98% |
| Chemical Name or Material | 2-Fluoro-6-nitrobenzyl bromide |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction