missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-6-methylpyridine, 97%, Thermo Scientific™
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| CAS | 407-22-7 |
| Boiling Point | 140°C to 141°C |
| Flash Point | 66°C |
| Assay Percent Range | 96% min. (GC) |
| Packaging | Glass Bottle |
| Molecular Formula | C6H6FN |
| MDL Number | MFCD00041226 |
| Quantity | 1 g |
| Synonym | 2-fluoro-6-picoline, 6-fluoro-2-picoline, 6-fluoro-2-methylpyridine, 6-fluoropicoline, pyridine, 2-fluoro-6-methyl, 2-fluoro-6-methyl-pyridine, 2-fluor-6-methylpyridin, pubchem6443, acmc-209jga |
| InChI Key | UDMNVTJFUISBFD-UHFFFAOYSA-N |
| Show More |
Safety and Handling
Product Identifier
- 2-Fluoro-6-methylpyridine
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction