missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxyphenol, 98%, Thermo Scientific™

Product Code. 10716455 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
This item is not returnable. View return policy

Product Code. 10716455

Brand: Thermo Scientific Acros 399950010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 73943-41-6
CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Brown
Density 1.2300g/mL
Boiling Point 130°C to 131°C (36.0 mmHg)
Flash Point 103°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C7H7FO2
Linear Formula FC6H3(OH)OCH3
Refractive Index 1.5160 to 1.5190 (20°C, 589nm)
MDL Number MFCD00075140
Quantity 1 g
Specific Gravity 1.23
Synonym 2-fluoro-6-methoxy-phenol, phenol, 2-fluoro-6-methoxy, pubchem3878, acmc-209osz, 2-methoxy-6-fluorophenol, 2-methoxyl-6-fluorophenol, 2-hydroxy-3-fluoroanisole, 2-fluoro-6-methoxy phenol, phenol,2-fluoro-6-methoxy, 2-fluoro-6-methoxyphenol
InChI Key YZNHPLVFLRSVHY-UHFFFAOYSA-N
SMILES COC1=C(O)C(F)=CC=C1
IUPAC Name 2-fluoro-6-methoxyphenol
Molecular Weight (g/mol) 142.13
PubChem CID 2737367
Formula Weight 142.13
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-6-methoxyphenol
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-6-methoxyphenol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.