missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxyphenol, 97%

Product Code. 11465820
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11465820

Brand: Thermo Scientific Alfa Aesar H26471.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 73943-41-6 | C7H7FO2 | 142.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73943-41-6
Molecular Formula C7H7FO2
Molecular Weight (g/mol) 142.13
MDL Number MFCD00075140
InChI Key YZNHPLVFLRSVHY-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxy-phenol, phenol, 2-fluoro-6-methoxy, pubchem3878, acmc-209osz, 2-methoxy-6-fluorophenol, 2-methoxyl-6-fluorophenol, 2-hydroxy-3-fluoroanisole, 2-fluoro-6-methoxy phenol, phenol,2-fluoro-6-methoxy, 2-fluoro-6-methoxyphenol
PubChem CID 2737367
IUPAC Name 2-fluoro-6-methoxyphenol
SMILES COC1=C(O)C(F)=CC=C1

Specifications

Density 1.23
Boiling Point 130°C to 131°C (36 mmHg)
Flash Point 103°C (218°F)
Refractive Index 1.52
Quantity 1 g
Formula Weight 142.13
Percent Purity 97%
Chemical Name or Material 2-Fluoro-6-methoxyphenol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.