missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzyl Bromide, TRC

Product Code. 30395387
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395387 100 mg
30403306 50 mg
30400429 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395387 Supplier Toronto Research Chemicals Supplier No. TRCF402810100MG
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500912-17-4
Molecular Formula C8H8OFBr
Molecular Weight (g/mol) 219.05
SMILES COc1cccc(F)c1CBr

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C8H8BrFO/c1-11-8-4-2-3-7(10)6(8)5-9/h2-4H,5H2,1H3
Formula Weight 217.9743
Chemical Name or Material 2-Fluoro-6-methoxybenzyl Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.