missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzyl bromide, 98%

Product Code. 15443407
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15443407 250 mg 250mg
11342139 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15443407 Supplier Thermo Scientific Alfa Aesar Supplier No. H26136.MD
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 500912-17-4 | C8H8BrFO | 219.053 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 500912-17-4
Molecular Formula C8H8BrFO
Molecular Weight (g/mol) 219.053
MDL Number MFCD08688579
InChI Key PVKFBCBTHHTDEX-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxybenzyl bromide, 2-bromomethyl-1-fluoro-3-methoxybenzene, 2-bromomethyl-3-fluoroanisole, 2-fluoro-6-methoxy-benzyl bromide, 2-bromomethyl-3-fluorophenyl methyl ether, benzene,2-bromomethyl-1-fluoro-3-methoxy, 2-bromomethyl-1-fluoranyl-3-methoxy-benzene
PubChem CID 2779322
IUPAC Name 2-(bromomethyl)-1-fluoro-3-methoxybenzene
SMILES COC1=C(C(=CC=C1)F)CBr

Specifications

Melting Point 18°C to 21°C
Refractive Index 1.557
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 219.05
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzyl bromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.