missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzoic acid, 97%, Thermo Scientific™

Product Code. 10097759 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10097759 5 g Glass bottle 5g
10037719 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10097759 Supplier Thermo Scientific Acros Supplier No. 436360050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 137654-21-8
Molecular Formula C8H7FO3
Molecular Weight (g/mol) 170.14
MDL Number MFCD00671764
InChI Key XLNQGZLCGVLNMF-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxy-benzoic acid, rarechem al bo 0442, benzoic acid, 2-fluoro-6-methoxy, pubchem4967, acmc-209cem, 6-fluoro-2-anisoic acid, intermediates-zcf02063, methyl-6-fluorosalicylic acid, ksc494o0r, 2-fluoro-6-methoxybenzoicacid
PubChem CID 2737360
IUPAC Name 2-fluoro-6-methoxybenzoic acid
SMILES COC1=C(C(=CC=C1)F)C(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 85°C to 87°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 170.14
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-methoxybenzoic acid
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-6-methoxybenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.