missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzeneboronic acid, 98%

Product Code. 11312009
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312009 5 g 5g
11302009 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312009 Supplier Thermo Scientific Alfa Aesar Supplier No. H26064.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 78495-63-3 | C7H8BFO3 | 169.946 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78495-63-3
Molecular Formula C7H8BFO3
Molecular Weight (g/mol) 169.946
MDL Number MFCD02179483
InChI Key XOVMDVZAWWQSDC-UHFFFAOYSA-N
Synonym 2-fluoro-6-methoxybenzeneboronic acid, 2-fluoro-6-methoxyphenyl boronic acid, 2-fluoro-6-methoxyphenylboronicacid, 6-fluoro-2-methoxyphenylboronic acid, 3-fluoroanisole-2-boronic acid, boronic acid, 2-fluoro-6-methoxyphenyl, 2-fluoro-6-methyloxyphenylboronic acid, boronic acid, b-2-fluoro-6-methoxyphenyl, 2-fluoro-6-methoxy-phenyl boronic acid, acmc-209pef
PubChem CID 3294524
IUPAC Name (2-fluoro-6-methoxyphenyl)boronic acid
SMILES B(C1=C(C=CC=C1F)OC)(O)O

Specifications

Melting Point 124°C to 126°C
Quantity 5 g
Formula Weight 169.95
Percent Purity 98%
Chemical Name or Material 2-Fluoro-6-methoxybenzeneboronic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.