missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-methoxybenzaldehyde, 98%, Thermo Scientific™

Product Code. 10442930 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10442930 1 g 1g
10658350 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10442930 Supplier Thermo Scientific Acros Supplier No. 435460010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 146137-74-8
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
MDL Number MFCD01090998
InChI Key UIOAYOIJMYMOEU-UHFFFAOYSA-N
Synonym 2-methoxy-6-fluorobenzaldehyde, 6-fluoro-o-anisaldehyde, 2-fluoro-6-methoxy-benzaldehyde, 2-fluoro-6-mehoxybenzaldehyde, benzaldehyde, 2-fluoro-6-methoxy, 2-fluoro-6-methoxybenzaldehyde, pubchem8486, acmc-209yud, 3-fluoro-2-formylanisole, intermediates-zcf02117
PubChem CID 2737357
IUPAC Name 2-fluoro-6-methoxybenzaldehyde
SMILES COC1=C(C=O)C(F)=CC=C1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 57.0°C to 60.0°C
Color Tan to White
Flash Point 110°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 154.14
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material 2-Fluoro-6-methoxybenzaldehyde
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-6-methoxybenzaldehyde
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.