missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodobenzoyl chloride, 97%, Thermo Scientific™

Product Code. 10224914 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
:
5g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10224914 5g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10224914 Supplier Thermo Scientific Acros Supplier No. 426130050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

CAS 111771-12-1
CAS Min % 96.0
CAS Max % 100.0
Density 1.9560g/mL
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C7H3ClFIO
Linear Formula FC6H3(I)COCl
MDL Number MFCD03094410
Quantity 5 g
Specific Gravity 1.956
Synonym 2-fluoro-6-iodobenzoylchloride, 2-fluoro-6-iodo-benzoyl chloride, pubchem4963, 2-fluoro-6-iodobenzoyl chloride
InChI Key KUULKUHCAMRGCF-UHFFFAOYSA-N
SMILES FC1=C(C(Cl)=O)C(I)=CC=C1
IUPAC Name 2-fluoro-6-iodobenzoyl chloride
Molecular Weight (g/mol) 284.45
PubChem CID 2778660
Formula Weight 284.45
Percent Purity 97%
Chemical Name or Material 2-Fluoro-6-iodobenzoyl chloride
Show More Show Less
compliance-icons
Product Identifier
  • 2-Fluoro-6-iodobenzoyl chloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • EUH029-Contact with water liberates toxic gas.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P402+P404-Store in a dry place. Store in a closed container.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.