missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-iodobenzoic acid, 98%, ACROS Organics™

Product Code. 10770581 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10770581 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10770581 Supplier Thermo Scientific Acros Supplier No. 311940010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 111771-08-5
CAS Min % 97.5
CAS Max % 100.0
Melting Point 121.0°C to 126.0°C
Color Yellow to Brown
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C7H4FIO2
Linear Formula FC6H3(I)CO2H
MDL Number MFCD00042289
Quantity 1 g
Synonym 6-fluoro-2-iodobenzoic acid, benzoic acid, 2-fluoro-6-iodo, maybridge3_007452, pubchem1386, acmc-2099an, 2-fluoro-6-iodobenzoicacid, 2-fluoro-6-iodo benzoic acid, 2-fluoro-6-iodo-benzoic acid, rarechem al bo 0280, attercop-chm at126295
InChI Key CYCXAPWOBWWNRK-UHFFFAOYSA-N
SMILES C1=CC(=C(C(=C1)I)C(=O)O)F
IUPAC Name 2-fluoro-6-iodobenzoic acid
Molecular Weight (g/mol) 266.01
PubChem CID 2733302
Formula Weight 266
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Fluoro-6-iodobenzoic acid, 98%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.