missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-6-hydroxyphenylboronic Acid, TRC

Product Code. 30395799
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395799 1 g
30397202 250 mg
30401953 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30395799 Supplier Toronto Research Chemicals Supplier No. TRCF4896601G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1256345-60-4
Molecular Formula C6H6BFO3
Molecular Weight (g/mol) 155.92
Synonym B-(2-Fluoro-6-hydroxyphenyl)boronic Acid
SMILES OB(O)c1c(O)cccc1F

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H6BFO3/c8-4-2-1-3-5(9)6(4)7(10)11/h1-3,9-11H
Formula Weight 156.0394
Chemical Name or Material 2-Fluoro-6-hydroxyphenylboronic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.