missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%

Product Code. 11369909
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11369909 1 g 1g
11379909 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11369909 Supplier Thermo Scientific Alfa Aesar Supplier No. B25630.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69922-27-6 | C8H3F4NO | 205.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69922-27-6
Molecular Formula C8H3F4NO
Molecular Weight (g/mol) 205.11
MDL Number MFCD00673069
InChI Key NAIKHCBDZGSGHH-UHFFFAOYSA-N
PubChem CID 2733380
IUPAC Name 1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1N=C=O)C(F)(F)F

Specifications

Density 1.418
Boiling Point 53°C (2.5 mmHg)
Flash Point 70°C (158°F)
Refractive Index 1.455
Quantity 1 g
UN Number UN2206
Sensitivity Moisture sensitive
Synonym 2-fluoro-5-trifluoromethyl phenyl isocyanate, 1-fluoro-2-isocyanato-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl phenylisocyanate, 2-fluoro-5-trifluoromethylphenyl isocyanate, benzene,1-fluoro-2-isocyanato-4-trifluoromethyl, benzene, 1-fluoro-2-isocyanato-4-trifluoromethyl, acmc-20anuz, pubchem12480, 2-fluoro-5-trifluoromethylphenylisocyanate, 2-fluoro-5-trifluoromethyl phenyl isocyan
Formula Weight 205.11
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenyl isocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.