missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific™
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass bottle
Specifications
CAS Min % | 96.0 |
CAS Max % | 100.0 |
Color | Colorless |
Boiling Point | 53°C (2.5 mmHg) |
Infrared Spectrum | Authentic |
Assay Percent Range | 97% |
Packaging | Glass bottle |
Linear Formula | FC6H3(CF3)NCO |
Refractive Index | 1.4540 to 1.456 |
Quantity | 2.5 g |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'