missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%, Thermo Scientific™

Product Code. 10126000 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
500 mg
2.5 g
Packaging:
Glass Bottle
Unit Size:
2.5g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10126000 500 mg Glass Bottle 500mg
10405115 2.5 g Glass Bottle 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10126000 Supplier Thermo Scientific Acros Supplier No. 319345000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 69922-27-6
Molecular Formula C8H3F4NO
Molecular Weight (g/mol) 205.11
MDL Number MFCD00673069
InChI Key NAIKHCBDZGSGHH-UHFFFAOYSA-N
PubChem CID 2733380
IUPAC Name 1-fluoro-2-isocyanato-4-(trifluoromethyl)benzene
SMILES FC1=CC=C(C=C1N=C=O)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Boiling Point 53°C (2.5 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Linear Formula FC6H3(CF3)NCO
Refractive Index 1.4540 to 1.456
Quantity 500 mg
Synonym 2-fluoro-5-trifluoromethyl phenyl isocyanate, 1-fluoro-2-isocyanato-4-trifluoromethyl benzene, 2-fluoro-5-trifluoromethyl phenylisocyanate, 2-fluoro-5-trifluoromethylphenyl isocyanate, benzene,1-fluoro-2-isocyanato-4-trifluoromethyl, benzene, 1-fluoro-2-isocyanato-4-trifluoromethyl, acmc-20anuz, pubchem12480, 2-fluoro-5-trifluoromethylphenylisocyanate, 2-fluoro-5-trifluoromethyl phenyl isocyan
Formula Weight 205.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)phenyl isocyanate, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.