missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethyl)anisole, 97%

Product Code. 11399809
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11399809 5 g 5g
11389809 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11399809 Supplier Thermo Scientific Alfa Aesar Supplier No. B23516.06
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 261951-78-4 | C8H6F4O | 194.129 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Not miscible in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 261951-78-4
Molecular Formula C8H6F4O
Molecular Weight (g/mol) 194.129
MDL Number MFCD01631564
InChI Key AXEAQZIYCQTHQV-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethyl anisole, 1-fluoro-2-methoxy-4-trifluoromethyl benzene, 4-fluoro-3-methoxybenzotrifluoride, pubchem8275, acmc-1ceet, 4-fluoro-3-methoxy-benzotrifluoride, 1-fluoranyl-2-methoxy-4-trifluoromethyl benzene
PubChem CID 2774771
IUPAC Name 1-fluoro-2-methoxy-4-(trifluoromethyl)benzene
SMILES COC1=C(C=CC(=C1)C(F)(F)F)F

Specifications

Flash Point 62°C (144°F)
Refractive Index 1.4352
Quantity 5 g
Solubility Information Not miscible in water.
Formula Weight 194.13
Percent Purity 97%
Chemical Name or Material 2-Fluoro-5-(trifluoromethyl)anisole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.