missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Fluoro-5-nitropyridine, 98%
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
Description
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
It is used in the preparation of nitropyridines by nitration of pyridines with nitric acid.
Solubility
Slightly soluble in water.
Notes
Moisture sensitive. Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas.
Specifications
Specifications
| Melting Point | 17°C to 21°C |
| Color | Colorless to Yellow |
| Density | 1.41 g/mL |
| Boiling Point | 86°C to 87°C (7 mmHg)(literature) |
| Refractive Index | 1.525 |
| Quantity | 5 g |
| Beilstein | 120351 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 142.09 |
| Show More |
Safety and Handling
Product Identifier
- 2-Fluoro-5-nitropyridine
- Warning
- Acute toxicity Category 4
- Specific target organ toxicity Category 3
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : 2-Fluoro-5-nitropyridine
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction